ENA333559477 Enamine

2-chloro-1-(1-methyl-1H-pyrazol-4-yl)ethan-1-one

  • MDL number MFCD10690121
  • Cas Number : 1152550-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84207
Availability details for
Quantity
  • Availability
  • Add to Cart