ENA333559487 Enamine

1,3-bis(4-methylphenyl)propan-2-one

  • MDL number MFCD11919450
  • Cas Number : 70769-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84234

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84234
Availability details for
Quantity
  • Availability
  • Add to Cart