ENA333559491 Enamine

6-{[4-(pyridin-4-yl)piperazin-1-yl]methyl}-1,3-dioxaindan-5-ol

  • MDL number MFCD13578203
  • Cas Number : 1197551-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83785
Availability details for
Quantity
  • Availability
  • Add to Cart