ENA333559493 Enamine

2-chloro-4-[3-(propan-2-yl)-1,2,4-oxadiazol-5-yl]pyridine

  • MDL number MFCD13881437
  • Cas Number : 1120245-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35...37
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84244

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84244
Availability details for
Quantity
  • Availability
  • Add to Cart