ENA333559494 Enamine

4-methyl-5-[4-(propan-2-yl)phenyl]thiophene-2-carboxylic acid

  • MDL number MFCD04971133
  • Cas Number : 832737-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83737
Availability details for
Quantity
  • Availability
  • Add to Cart