ENA333559501 Enamine

4-methyl-5-(4-methylphenyl)thiophene-2-carboxylic acid

  • MDL number MFCD04971174
  • Cas Number : 832737-68-5
  • Molecular Weight : 232.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-83738

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83738
Availability details for
Quantity
  • Availability
  • Add to Cart