ENA333559508 Enamine

3-(3,4-dihydro-2H-1,5-benzodioxepine-7-sulfonyl)propanoic acid

  • MDL number MFCD09047648
  • Cas Number : 926204-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-84299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84299
Availability details for
Quantity
  • Availability
  • Add to Cart