ENA333559517 Enamine

1-(2-methoxyethoxy)propan-2-one

  • MDL number MFCD01105045
  • Cas Number : 88986-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84318
Availability details for
Quantity
  • Availability
  • Add to Cart