ENA333559522 Enamine

N-(3,4-dichlorophenyl)piperidine-4-carboxamide

  • MDL number MFCD09808990
  • Cas Number : 894802-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84324
Availability details for
Quantity
  • Availability
  • Add to Cart