ENA333559523 Enamine

6-chloro-N-methylpyridazine-3-carboxamide

  • MDL number MFCD12165803
  • Cas Number : 202135-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84327

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-84327
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart