ENA333559526 Enamine

5-chloro-1,3-thiazol-2-amine

  • MDL number MFCD01681563
  • Cas Number : 41663-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90...92
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84362

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-84362
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart