ENA333559533 Enamine

N-carbamimidoylformamide

  • MDL number MFCD00021034
  • Cas Number : 4471-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-180
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84390
Availability details for
Quantity
  • Availability
  • Add to Cart