ENA333559544 Enamine

3,4-diaminobutan-1-ol

  • MDL number MFCD19206877
  • Cas Number : 98021-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84444
Availability details for
Quantity
  • Availability
  • Add to Cart