ENA333559550 Enamine

methyl 6-bromo-4-chloroquinoline-2-carboxylate

  • MDL number MFCD07783793
  • Cas Number : 887587-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-83826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83826
Availability details for
Quantity
  • Availability
  • Add to Cart