ENA333559553 Enamine

3-ethoxy-N-methylspiro[3.4]octan-1-amine hydrochloride

  • MDL number MFCD19982541
  • Cas Number : 1333616-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168...170
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83829

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83829
Availability details for
Quantity
  • Availability
  • Add to Cart