ENA333559555 Enamine

5-fluoro-2-(morpholin-4-yl)benzonitrile

  • MDL number MFCD11037711
  • Cas Number : 905439-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-83832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83832
Availability details for
Quantity
  • Availability
  • Add to Cart