ENA333559566 Enamine

1,3-dithian-5-one

  • MDL number MFCD19982619
  • Cas Number : 64211-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-105
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84644
Availability details for
Quantity
  • Availability
  • Add to Cart