ENA333559581 Enamine

3-fluoropropane-1-sulfonyl fluoride

  • MDL number MFCD01697638
  • Cas Number : 461-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84685
Availability details for
Quantity
  • Availability
  • Add to Cart