ENA333559582 Enamine

3-fluoropropane-1-sulfonyl chloride

  • MDL number MFCD01697637
  • Cas Number : 461-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84687

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-84687
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart