ENA333559584 Enamine

3-fluoropropan-1-amine

  • MDL number MFCD01694865
  • Cas Number : 462-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Flash Point 21
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84693

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-84693
pictograms GHS02,GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart