ENA336955632 Enamine

[4-(ethylsulfanyl)oxan-4-yl]methanamine

  • MDL number MFCD20233442
  • Cas Number : 1354952-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-74712

Safety Information

msds https://www.enaminestore.com/catalog/EN300-74712
Availability details for
Quantity
  • Availability
  • Add to Cart