ENA336955636 Enamine

1,3-benzothiazole-5-sulfonyl chloride

  • MDL number MFCD20233448
  • Cas Number : 227278-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-78162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-78162
Availability details for
Quantity
  • Availability
  • Add to Cart