ENA336955637 Enamine

4-chloro-6-(chloromethyl)-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD20233449
  • Cas Number : 1354952-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-79464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-79464
Availability details for
Quantity
  • Availability
  • Add to Cart