ENA336955643 Enamine

N-(4-aminocyclohexyl)methanesulfonamide hydrochloride

  • MDL number MFCD20233459
  • Cas Number : 1206776-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-82998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-82998
Availability details for
Quantity
  • Availability
  • Add to Cart