ENA336955655 Enamine

decahydro-1H-pyrrolo[3,4-a]indolizine

  • MDL number MFCD20233478
  • Cas Number : 1354954-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-84305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84305
Availability details for
Quantity
  • Availability
  • Add to Cart