ENA336955656 Enamine

3-[(5-chloro-2-methoxyphenyl)methyl]piperidine hydrochloride

  • MDL number MFCD20233480
  • Cas Number : 1354951-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-84322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84322
Availability details for
Quantity
  • Availability
  • Add to Cart