ENA336955667 Enamine

7-(methylsulfanyl)-2,3-dihydro-1,4-benzodioxine-6-carbaldehyde

  • MDL number MFCD20233507
  • Cas Number : 1354951-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84703

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84703
Availability details for
Quantity
  • Availability
  • Add to Cart