ENA336955690 Enamine

5-cyclohexyl-N-methyl-1H-pyrazol-3-amine

  • MDL number MFCD20233534
  • Cas Number : 1354949-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84777
Availability details for
Quantity
  • Availability
  • Add to Cart