ENA336955695 Enamine

2-[1-(methylamino)cyclohexyl]acetic acid hydrochloride

  • MDL number MFCD20233539
  • Cas Number : 1354950-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84817
Availability details for
Quantity
  • Availability
  • Add to Cart