ENA336955697 Enamine

(4,5-dimethylthiophen-2-yl)methanamine hydrochloride

  • MDL number MFCD20233544
  • Cas Number : 1354962-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84846
Availability details for
Quantity
  • Availability
  • Add to Cart