ENA336955703 Enamine

4-isocyanatocyclopent-1-ene

  • MDL number MFCD20233552
  • Cas Number : 2487-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84912

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84912
Availability details for
Quantity
  • Availability
  • Add to Cart