ENA336955706 Enamine

N-methyl-2-[2-(methylamino)ethoxy]acetamide hydrochloride

  • MDL number MFCD20233555
  • Cas Number : 1354953-86-8
  • Molecular Weight : 182.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84958
Availability details for
Quantity
  • Availability
  • Add to Cart