ENA336955714 Enamine

2-(4-hydroxy-1-methylpiperidin-4-yl)acetic acid hydrochloride

  • MDL number MFCD20233562
  • Cas Number : 1354953-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85046

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85046
Availability details for
Quantity
  • Availability
  • Add to Cart