ENA336955728 Enamine

3-(oxan-4-yloxy)propan-1-amine hydrochloride

  • MDL number MFCD20233576
  • Cas Number : 1354953-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85137
Availability details for
Quantity
  • Availability
  • Add to Cart