ENA336955729 Enamine

2-methyl-2-(2-methylphenyl)cyclopropan-1-amine hydrochloride

  • MDL number MFCD20233577
  • Cas Number : 1354951-37-3
  • Molecular Weight : 197.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-2-(2-methylphenyl)cyclopropanamine hydrochloride
Inchi Code 1S/C11H15N.ClH/c1-8-5-3-4-6-9(8)11(2)7-10(11)12;/h3-6,10H,7,12H2,1-2H3;1H
Inchi Key WLDUEGOJHABMOI-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-85138

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-85138
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart