ENA336955734 Enamine

2-(2-methoxyphenyl)-2-methylcyclopropan-1-amine hydrochloride

  • MDL number MFCD20233582
  • Cas Number : 1354954-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85146
Availability details for
Quantity
  • Availability
  • Add to Cart