ENA336955742 Enamine

1-[2-(chloromethyl)phenyl]pyrrolidine hydrochloride

  • MDL number MFCD20233591
  • Cas Number : 110405-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85343
Availability details for
Quantity
  • Availability
  • Add to Cart