ENA336955743 Enamine

2-(2,6-difluorophenyl)-2-methylcyclopropan-1-amine hydrochloride

  • MDL number MFCD20233592
  • Cas Number : 1354954-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85345
Availability details for
Quantity
  • Availability
  • Add to Cart