ENA336955755 Enamine

(2-chlorophenyl)(3-methoxyphenyl)methanamine hydrochloride

  • MDL number MFCD20233606
  • Cas Number : 1354954-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-85442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85442
Availability details for
Quantity
  • Availability
  • Add to Cart