ENA336955772 Enamine

3-[3-(1-aminoethyl)phenyl]-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD20233624
  • Cas Number : 1354962-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85540
Availability details for
Quantity
  • Availability
  • Add to Cart