ENA336955786 Enamine

4-(methylamino)-1lambda4-thian-1-one hydrochloride

  • MDL number MFCD20233639
  • Cas Number : 1354951-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85672

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85672
Availability details for
Quantity
  • Availability
  • Add to Cart