ENA336955795 Enamine

2-(2-aminobutan-2-yl)-6-(methoxymethyl)-3,4-dihydropyrimidin-4-one hydrochloride

  • MDL number MFCD20233652
  • Cas Number : 1354950-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85875
Availability details for
Quantity
  • Availability
  • Add to Cart