ENA336955797 Enamine

3-(4-butylcyclohexyl)-1H-pyrazol-5-amine hydrochloride

  • MDL number MFCD20233657
  • Cas Number : 1486498-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-85944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85944
Availability details for
Quantity
  • Availability
  • Add to Cart