ENA336955809 Enamine

[2-(3-methanesulfonylphenoxy)pyridin-3-yl]methanamine

  • MDL number MFCD12536314
  • Cas Number : 1157379-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84281

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84281
Availability details for
Quantity
  • Availability
  • Add to Cart