ENA336955812 Enamine

4-(aminomethyl)-4-hydroxy-1lambda6-thiane-1,1-dione

  • MDL number MFCD16045747
  • Cas Number : 858823-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84303
Availability details for
Quantity
  • Availability
  • Add to Cart