ENA336955814 Enamine

2-(3-methylphenyl)propan-1-amine

  • MDL number MFCD14622450
  • Cas Number : 5266-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84306

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84306
Availability details for
Quantity
  • Availability
  • Add to Cart