ENA336955815 Enamine

7,7-dimethyl-2-oxabicyclo[3.2.0]heptan-6-amine

  • MDL number MFCD20233479
  • Cas Number : 1341434-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84307

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84307
Availability details for
Quantity
  • Availability
  • Add to Cart