ENA336955819 Enamine

4-methanesulfinylaniline

  • MDL number MFCD14631763
  • Cas Number : 22865-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-51157

Safety Information

msds https://www.enaminestore.com/catalog/EN300-51157
Availability details for
Quantity
  • Availability
  • Add to Cart