ENA336955822 Enamine

1-(2-methanesulfinylethyl)piperazine dihydrochloride

  • MDL number MFCD18071280
  • Cas Number : 1332528-51-4
  • Molecular Weight : 249.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84353
Availability details for
Quantity
  • Availability
  • Add to Cart