ENA336955826 Enamine

3-(cyclopentyloxy)-4-methylaniline

  • MDL number MFCD11935260
  • Cas Number : 1154943-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-84357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84357
Availability details for
Quantity
  • Availability
  • Add to Cart